EMAIL THIS PAGE TO A FRIEND
16115 Sigma-Aldrich

Bromoacetyl bromide

purum, ≥98.0% (AT)

  • CAS Number 598-21-0

  • Linear Formula BrCH2COBr

  • Molecular Weight 201.84

  •  Beilstein Registry Number 605440

  •  EC Number 209-923-5

  •  MDL number MFCD00000115

  •  PubChem Substance ID 24849908

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
vapor pressure   3.8 mmHg ( 25 °C)
InChI Key   LSTRKXWIZZZYAS-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
refractive index   n20/D 1.547(lit.)
  n20/D 1.547
bp   147-150 °C(lit.)
density   2.324 g/mL at 20 °C
  2.317 g/mL at 25 °C(lit.)

Description

Caution

discolor to brown on storage

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2513 8 / PGII
WGK Germany 
3
Flash Point(F) 
221 °F
Flash Point(C) 
105 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?