161519 Sigma-Aldrich

Sodium metabisulfite

reagent grade, 97%

DOWNLOAD MSDS (PDF)

Synonym: Sodium disulfite, Sodium pyrosulfite

Properties

Related Categories Essential Chemicals, Inorganic Salts, Solutions and Reagents, Reagent Grade, Research Essentials
grade   reagent grade
assay   97%
mp   >300 °C(lit.)

Description

Packaging

2.5 kg in poly bottle

500 g in poly bottle

Price and Availability

Customers Also Viewed

Sodium bisulfite

ACS reagent, mixture of NaHSO3 and Na2S2O5

Sodium metabisulfite

puriss. p.a., ACS reagent, reag. Ph. Eur., dry, 98-100.5%

Sodium metabisulfite

puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5%

Sodium metabisulfite

ACS reagent, ≥97.0%

Sodium metabisulfite

ReagentPlus®, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?