• USA Home
  • 161527 - Sodium hydrosulfide hydrate

EMAIL THIS PAGE TO A FRIEND

161527 Sigma-Aldrich

Sodium hydrosulfide hydrate

Purchase

Description

Frequently Asked Questions

Live Chat and Frequently Asked Questions are available for this Product.

Packaging

1 kg in glass bottle

3 kg in steel drum

5, 100 g in glass bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Ultra-high Purity Salts

Additional Products to Explore

Customers Also Viewed

Hydrogen sulfide

purum, ≥99.0%

Hydrogen sulfide

≥99.5%

Sodium hydrosulfide monohydrate

technical, ≥90%

Sodium sulfide hydrate

≥60%, scales

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2949 8 / PGII
WGK Germany 
2
Flash Point(F) 
194 °F
Flash Point(C) 
90 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Fieser 3,266 / Fieser 11,487

Corp MSDS 1 (2), 3143:C / IR-Spectra (2), 1339:F / IR-Spectra (3), 1567:F / RegBook 1 (3), 3295:C

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?