EMAIL THIS PAGE TO A FRIEND
16167 Sigma-Aldrich

SODOSIL RM 02

neutral

  •  UBA Number 08050025

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents N to Z, Non-Ionic Detergents
pH   8-10 (20 °C, direct)
density   1.04-1.10 g/mL at 20 °C

Description

Reconstitution

The best cleaning results are achieved with = 2−5% for normal contamination. The cleaning time = 2−24 h depends on the degree of contamination and on the bath temperature. The action of ultra-sonic waves accelerates the cleaning operation considerably as does an increase in temperature. The cleaning solution can be prepared with tap water. The use of demineralisied water results in more optimum utilisation of baths.

Legal Information

SODOSIL is a trademark of Sigma-Aldrich International GmbH

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?