EMAIL THIS PAGE TO A FRIEND
16168 Sigma-Aldrich

SODOSIL RM 03

phosphate-free cleaner

  •  UBA Number 08050026

Purchase

Properties

pH   12-14 (20 °C, direct)

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Reconstitution

The best cleaning results are achieved with = 2−20% for normal contamination. The cleaning time = 2−24 h depends on the degree of contamination and on the bath temperature. The action of ultra-sonic waves accelerates the cleaning operation considerably as does an increase in temperature. The cleaning solution can be prepared with tap water. The use of demineralisied water results in more optimum utilisation of baths.

Legal Information

SODOSIL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3267 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?