EMAIL THIS PAGE TO A FRIEND
16181 Sigma-Aldrich

SODOSIL RA 31

anti-foaming agent

  •  UBA Number 08050019

Purchase

Properties

Related Categories Cleaners, Detergents, Labware More...
impurities   ≥30% polydimethylsiloxane
pH   6.5

Description

Legal Information

SODOSIL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?