EMAIL THIS PAGE TO A FRIEND
16182 Sigma-Aldrich

SODOSIL RA 32

clear rinse

  •  EC Number 203-473-3

  •  UBA Number 08050032

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents N to Z, Non-Ionic Detergents
description   non-ionic
composition   nonionic surfactants, 5-15%
pH   1.5-1.7

Description

Legal Information

SODOSIL is a trademark of Sigma-Aldrich International GmbH

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
1
Flash Point(F) 
231.8 °F
Flash Point(C) 
111 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?