EMAIL THIS PAGE TO A FRIEND
16325 Sigma-Aldrich

D-(+)-Glucose

meets analytical specification of Ph. Eur., BP, anhydrous

Synonym: Dextrose

  • CAS Number 50-99-7

  • Empirical Formula (Hill Notation) C6H12O6

  • Molecular Weight 180.16

  •  Beilstein Registry Number 1724615

  •  EC Number 200-075-1

  •  MDL number MFCD00063774

  •  PubChem Substance ID 24850031

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Carbohydrates, Chemical Biology, Chemical Synthesis,
grade   anhydrous
  meets analytical specification of Ph. Eur., BP
InChI Key   WQZGKKKJIJFFOK-DVKNGEFBSA-N
optical activity   [α]20/D +52.5 to +53.3°, c = 10% in H2O (calc. on anhydrous subst.)
quality   meets analytical specification of Ph. Eur., BP
impurities   acidity or alkalinity, complies
  dextrine, complies
  foreign sugar, complies
  residual solvents, complies
  soluble starch, complies
  ≤0.001% sulfite (SO2)
  ≤1% water (Karl Fischer)
ign. residue   ≤0.1% (as SO4)
mp   150-152 °C(lit.)
solubility   H2O: soluble
anion traces   chloride (Cl-): ≤100 mg/kg
  sulfate (SO42-): ≤200 mg/kg
cation traces   As: ≤1 mg/kg
  Ca: ≤200 mg/kg
  Pb: ≤0.5 mg/kg
suitability   complies for appearance of solution
Gene Information   human ... PYGM(5837)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
LZ6600000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?