• USA Home
  • 16370 - 4-Bromobenzenesulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
16370 Sigma-Aldrich

4-Bromobenzenesulfonyl chloride

purum, ≥98.0% (AT)

  • CAS Number 98-58-8

  • Linear Formula BrC6H4SO2Cl

  • Molecular Weight 255.52

  •  Beilstein Registry Number 743518

  •  EC Number 202-683-2

  •  MDL number MFCD00007437

  •  PubChem Substance ID 329751200

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonyl Halides, Sulfur Compounds More...
InChI Key   KMMHZIBWCXYAAH-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
bp   153 °C/15 mmHg(lit.)
mp   73-75 °C(lit.)
  73-75 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?