EMAIL THIS PAGE TO A FRIEND
16495 Sigma-Aldrich

2-Bromo-2-butene

purum, cis+trans, ≥97.0% (GC)

  • CAS Number 13294-71-8

  • Linear Formula CH3CH=C(Br)CH3

  • Molecular Weight 135.00

  •  Beilstein Registry Number 1733329

  •  EC Number 236-316-2

  •  MDL number MFCD00000141

Purchase

Properties

InChI Key   UILZQFGKPHAAOU-ONEGZZNKSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.459
bp   86-87 °C(lit.)
density   1.320 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BF 3 / PGII
WGK Germany 
3
Flash Point(F) 
33.8 °F
Flash Point(C) 
1 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

215562 2-Bromo-2-butene, mixture of cis and trans, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?