• USA Home
  • 16515 - 4-Bromobutyryl chloride

EMAIL THIS PAGE TO A FRIEND
16515 Sigma-Aldrich

4-Bromobutyryl chloride

purum, ≥98.0% (GC)

  • CAS Number 927-58-2

  • Linear Formula BrCH2CH2CH2COCl

  • Molecular Weight 185.45

  •  Beilstein Registry Number 1743026

  •  EC Number 213-154-0

  •  MDL number MFCD00000753

  •  PubChem Substance ID 329751257

Purchase

Properties

Related Categories Acid Halides, Bifunctional Linkers, Building Blocks, Carbonyl Compounds, Chemical Biology,
InChI Key   LRTRXDSAJLSRTG-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.492(lit.)
  n20/D 1.493
bp   101 °C/37 mmHg(lit.)
density   1.60 g/mL at 20 °C
  1.602 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
195.8 °F
Flash Point(C) 
91 °C
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

251933 4-Bromobutyryl chloride, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?