EMAIL THIS PAGE TO A FRIEND
16594 Sigma-Aldrich

6-Bromohexanoic acid

purum, ≥97.0% (AT)

Synonym: 6-Bromocaproic acid

  • CAS Number 4224-70-8

  • Linear Formula BrCH2(CH2)4COOH

  • Molecular Weight 195.05

  •  Beilstein Registry Number 1749740

  •  EC Number 224-176-5

  •  MDL number MFCD00004422

  •  PubChem Substance ID 24850150

Purchase

Properties

Related Categories Building Blocks, C6, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   NVRVNSHHLPQGCU-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
bp   165-170 °C/20 mmHg(lit.)
mp   32-34 °C(lit.)
solubility   methanol: soluble0.1 g/mL, clear, colorless to faintly brownish-yellow

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
Flash Point(F) 
203 °F
Flash Point(C) 
95 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

150452 6-Bromohexanoic acid, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?