• USA Home
  • 16663 - 1-Bromo-6-chlorohexane

EMAIL THIS PAGE TO A FRIEND
16663 Sigma-Aldrich

1-Bromo-6-chlorohexane

purum, ≥95.0% (GC)

  • CAS Number 6294-17-3

  • Linear Formula Br(CH2)6Cl

  • Molecular Weight 199.52

  •  Beilstein Registry Number 1697304

  •  EC Number 228-555-6

  •  MDL number MFCD00001019

Purchase

Properties

InChI Key   JTYUIAOHIYZBPB-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.481(lit.)
  n20/D 1.482
bp   109-110 °C/2 mmHg(lit.)
density   1.337 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

1-Bromo-6-chlorohexane was used in the synthesis of 4-(6-chlorohexoxy)benzophenone.

General description

1-Bromo-6-chlorohexane reacts with nBuMgCl in the presence of CuCl2 and 1-phenylpropyne at 0°C to yield 2,2-dimethyldodecane.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3
Flash Point(F) 
213.8 °F
Flash Point(C) 
101 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

241652 1-Bromo-6-chlorohexane, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?