• USA Home
  • 16730 - 2-Bromo-2-chloro-1,1,1-trifluoroethane

EMAIL THIS PAGE TO A FRIEND
16730 Sigma-Aldrich

2-Bromo-2-chloro-1,1,1-trifluoroethane

≥99.0% (GC)

Synonym: Halothane

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
vapor pressure   4.5 psi ( 20 °C)
InChI Key   BCQZXOMGPXTTIC-UHFFFAOYSA-N
assay   ≥99.0% (GC)
refractive index   n20/D 1.369(lit.)
  n20/D 1.369
bp   50.2 °C(lit.)
density   1.872 g/mL at 25 °C(lit.)
storage temp.   2-8°C
Gene Information   human ... ATP5D(513), GLRA1(2741), GRIA1(2890), KCNN4(3783)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Biochem/physiol Actions

Inhalation anesthetic. In an effect believed to be independent of anesthesia, halothane inhibits synthesis of 5-hydroxytryptamine in brain tissue, probably at the tryptophan hydroxylase step.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3334
WGK Germany 
3
RTECS 
KH6550000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?