EMAIL THIS PAGE TO A FRIEND
16810 Sigma-Aldrich

Bromocyclohexane

purum, ≥99.0% (GC)

Synonym: Cyclohexyl bromide

  • CAS Number 108-85-0

  • Empirical Formula (Hill Notation) C6H11Br

  • Molecular Weight 163.06

  •  Beilstein Registry Number 1098373

  •  EC Number 203-622-2

  •  MDL number MFCD00003819

  •  PubChem Substance ID 329751385

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   AQNQQHJNRPDOQV-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.495(lit.)
  n20/D 1.495
bp   166-167 °C(lit.)
density   1.324 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
145.4 °F
Flash Point(C) 
63 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

135194 Bromocyclohexane, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?