EMAIL THIS PAGE TO A FRIEND
16845 Sigma-Aldrich

5-Bromocytosine

purum, ≥95.0% (NT)

Synonym: 4-Amino-5-bromo-2-hydroxypyrimidine

  • CAS Number 2240-25-7

  • Empirical Formula (Hill Notation) C4H4BrN3O

  • Molecular Weight 190.00

  •  Beilstein Registry Number 127286

  •  EC Number 218-806-8

  •  MDL number MFCD00056025

  •  PubChem Substance ID 329751396

Purchase

Properties

InChI Key   QFVKLKDEXOWFSL-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (NT)
mp   240-243 °C (dec.)(lit.)
  ~250 °C (dec.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

150649 5-Bromocytosine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?