EMAIL THIS PAGE TO A FRIEND
16850 Sigma-Aldrich

1-Bromodecane

purum, ≥97.0% (GC)

Synonym: Decyl bromide

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   MYMSJFSOOQERIO-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.456(lit.)
  n20/D 1.456
bp   238 °C(lit.)
density   1.066 g/mL at 25 °C(lit.)

Description

Other Notes

This product has been replaced by 145785-ALDRICH | 1-Bromodecane 98%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN3082 - class 9 - PG 3 - DOT NA1993 - Environmentally hazardous substances, liquid, n.o.s. HI: all (not BR)
WGK Germany 
3
RTECS 
HD6850000
Flash Point(F) 
201.2 °F
Flash Point(C) 
94 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

145785 1-Bromodecane, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?