• USA Home
  • 16880 - 5-Bromo-2′-deoxyuridine

EMAIL THIS PAGE TO A FRIEND
16880 Sigma-Aldrich

5-Bromo-2′-deoxyuridine

≥99.0% (HPLC)

Synonym: 5-BrdU, 5-Bromo-1-(2-deoxy-β-D-ribofuranosyl)uracil, 5-Bromouracil deoxyriboside, BUdR

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   WOVKYSAHUYNSMH-RRKCRQDMSA-N
assay   ≥99.0% (HPLC)
optical activity   [α]20/D +23±1°, c = 1% in H2O
mp   191-194 °C (dec.)(lit.)
storage temp.   2-8°C

Description

Biochem/physiol Actions

Thymidine analog used as a mutagen in genetic research. Selectively incorporated into cellular DNA during S-phase.

Other Notes

Thymidine analogue. Inhibitor of cell differentiation : Review; Incorporation into DNA for flow cytometric analysis

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
2
RTECS 
YU7350000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?