EMAIL THIS PAGE TO A FRIEND
16895 Sigma-Aldrich

Bromodichloromethane

purum, ≥98.0% (GC), contains silver wool as stabilizer

Synonym: Dichlorobromomethane

  • CAS Number 75-27-4

  • Linear Formula BrCHCl2

  • Molecular Weight 163.83

  •  Beilstein Registry Number 1697005

  •  EC Number 200-856-7

  •  MDL number MFCD00000824

  •  PubChem Substance ID 24850257

Purchase

Properties

InChI Key   FMWLUWPQPKEARP-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
contains   silver wool as stabilizer
refractive index   n20/D 1.497(lit.)
  n20/D 1.498
bp   87 °C(lit.)
mp   −55 °C(lit.)
density   1.98 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
PA5310000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?