EMAIL THIS PAGE TO A FRIEND
16987 Sigma-Aldrich

1-Bromoeicosane

purum, ≥97.0% (GC)

Synonym: Eicosyl bromide

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
vapor density   12.5 (vs air)
vapor pressure   <1 mmHg ( 20 °C)
InChI Key   CZASMUMJSKOHFJ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
mp   36-39 °C(lit.)
  36-39 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?