EMAIL THIS PAGE TO A FRIEND
17010 Sigma-Aldrich

Bromoacetic acid

purum, ≥98.0% (T)

  • CAS Number 79-08-3

  • Linear Formula BrCH2COOH

  • Molecular Weight 138.95

  •  Beilstein Registry Number 506167

  •  EC Number 201-175-8

  •  MDL number MFCD00002678

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Organic Acids, Synthetic Reagents More...
InChI Key   KDPAWGWELVVRCH-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
impurities   ≤2% dibromoacetic acid (1H-NMR)
bp   208 °C(lit.)
mp   44-49 °C
  47-49 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3425 8 / PGII
WGK Germany 
2
RTECS 
AF5950000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?