EMAIL THIS PAGE TO A FRIEND
17023 Sigma-Aldrich

Ethyl acetate

tested according to Ph.Eur.

Synonym: EtOAc, Ethylis acetas

Purchase

Properties

Related Categories Analytical/Chromatography, Ethyl Acetate, Pharmacopoeia, Pharmacopoeia A-Z, Solvent by Type,
grade   Ph Eur (tested according to)
vapor density   3 (20 °C, vs air)
vapor pressure   73 mmHg ( 20 °C)
InChI Key   XEKOWRVHYACXOJ-UHFFFAOYSA-N
product line   SAFC Hitech®
autoignition temp.   801 °F
pharmacopeia   testing conforms to Pharmacopeia
shelf life   02 yr
expl. lim.   2.2-11.5 %, 38 °F
refractive index   n20/D 1.3720(lit.)
bp   76.5-77.5 °C(lit.)
mp   −84 °C(lit.)
solubility   water: soluble
density   0.902 g/mL at 25 °C(lit.)

Description

Application

Ethyl acetate is a food odorant used as a test reagent for determining the olfactory rhythms in Drosophila melanogaster by measuring electroantennogram (EAG) responses to odorants.

Biochem/physiol Actions

Ethyl acetate is the ester of ethanol and acetic acid that possess antioxidant property and can induce genotoxicity in V79 mammalian cells.

Legal Information

SAFC Hitech is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1173 - class 3 - PG 2 - Ethyl acetate
WGK Germany 
1
RTECS 
AH5425000
Flash Point(F) 
26.6 °F
Flash Point(C) 
-3 °C
Protocols & Articles

Related Content

Properties of Solvents Table

a. Gant, R., LC/GC 10(7), 515 (1992) 1. At 20 °C 2. At 20 °C +/- 5 °C relative to water at 4 °C 3. At 10mmHg pressure 4. At l=589.32nm
Keywords: Gas chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?