• USA Home
  • 17035 - tert-Butyl bromoacetate

EMAIL THIS PAGE TO A FRIEND
17035 Sigma-Aldrich

tert-Butyl bromoacetate

purum, ≥97.0% (GC)

  • CAS Number 5292-43-3

  • Linear Formula BrCH2COOC(CH3)3

  • Molecular Weight 195.05

  •  Beilstein Registry Number 1753010

  •  EC Number 226-133-6

  •  MDL number MFCD00000188

  •  PubChem Substance ID 24850297

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   BNWCETAHAJSBFG-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.445(lit.)
  n20/D 1.445
bp   50 °C/10 mmHg(lit.)
density   1.321 g/mL at 25 °C(lit.)

Description

Other Notes

Reagent used in Reformatzky reactions; Alkylation of Reformatzky reagents

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
120.2 °F
Flash Point(C) 
49 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?