• USA Home
  • 17038 - Sodium standard for ICP

EMAIL THIS PAGE TO A FRIEND
17038 Sigma-Aldrich

Sodium standard for ICP

for ICP, ready-to-use, in nitric acid

Synonym: Sodium nitrate solution

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, ICP-OES/-MS, Single Solution,
grade   for ICP
InChI Key   KEAYESYHFKHZAL-UHFFFAOYSA-N
CofA   the measured values of the metal traces are reported on Certificate of Analysis
shelf life   limited shelf life, expiry date on the label
concentration   in nitric acid
  1.000 g/L Na
application(s)   ICP/AAS: suitable
format   single component solution

Description

Preparation Note

prepared with NaNO3 and HNO3 (~0.5 M)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

00462 Sodium Standard for ICP, TraceCERT®, 1000 mg/L Na in nitric acid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?