• USA Home
  • 17051 - Thallium standard for ICP

EMAIL THIS PAGE TO A FRIEND
17051 Sigma-Aldrich

Thallium standard for ICP

for ICP, ready-to-use, in nitric acid

Synonym: Thallium(I) nitrate solution

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, ICP-OES/-MS, Single Solution,
grade   for ICP
InChI Key   BKVIYDNLLOSFOA-UHFFFAOYSA-N
CofA   the measured values of the metal traces are reported on Certificate of Analysis
shelf life   limited shelf life, expiry date on the label
concentration   in nitric acid
  1.000 g/L Tl
application(s)   ICP/AAS: suitable
format   single component solution

Description

Preparation Note

prepared with TlNO3 and HNO3 (~0.5 M)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

51873 Thallium Standard for ICP, TraceCERT®, 1000 mg/L Tl in nitric acid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?