• USA Home
  • 17056 - Barium standard for ICP

EMAIL THIS PAGE TO A FRIEND
17056 Sigma-Aldrich

Barium standard for ICP

for ICP, ready-to-use, traceable to BAM, in H2O

Synonym: Barium nitrate solution

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Barium, Chemical Synthesis,
grade   for ICP
InChI Key   DSAJWYNOEDNPEQ-UHFFFAOYSA-N
CofA   the measured values of the metal traces are reported on Certificate of Analysis
shelf life   limited shelf life, expiry date on the label
concentration   in H2O
  1.000 g/L Ba
application(s)   ICP/AAS: suitable
format   single component solution

Description

Preparation Note

prepared with Ba(NO3)2 and H2O

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?