• USA Home
  • 17060 - 1-Bromo-2-fluorobenzene

EMAIL THIS PAGE TO A FRIEND
17060 Sigma-Aldrich

1-Bromo-2-fluorobenzene

purum, ≥98.0% (GC)

  • CAS Number 1072-85-1

  • Linear Formula BrC6H4F

  • Molecular Weight 175.00

  •  Beilstein Registry Number 1071819

  •  EC Number 214-018-3

  •  MDL number MFCD00000282

Purchase

Properties

Related Categories Aryl, Aryl Fluorinated Building Blocks, Building Blocks, C6, Chemical Synthesis,
InChI Key   IPWBFGUBXWMIPR-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.534(lit.)
  n20/D 1.534
bp   78-82 °C/50 mmHg
density   1.601 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

1-Bromo-2-fluorobenzene was used in the preparation of N-(dimethylaminoethyl)-2-fluoroaniline.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
RTECS 
DA1050000
Flash Point(F) 
118.4 °F
Flash Point(C) 
48 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?