EMAIL THIS PAGE TO A FRIEND
17079 Sigma-Aldrich

Fluo-3FF-AM

for fluorescence, ≥95% (TLC)

  • Empirical Formula (Hill Notation) C50H46Cl2F2N2O23

  • Molecular Weight 1151.80

  •  MDL number MFCD01866560

  •  PubChem Substance ID 329751471

Purchase

Properties

Related Categories Biochemicals and Reagents, Calcium Indicators, Chelators and Ion Probes, Fluorescent Indicators and Probes, Fluorescent Probes, Labels, Particles and Stains,
grade   for fluorescence
InChI Key   ABZBPPQDKRLSCZ-UHFFFAOYSA-N
assay   ≥95% (TLC)
solubility   DMSO: soluble
fluorescence   λex 500 nm; λem 526 nm in 0.1 M Tris pH 8.0, esterase; 10 mM Ca2+
storage temp.   −20°C

Description

Features and Benefits

Red solid. Cell-permeable acetoxymethyl ester (AM) derivative of Fluo-3FF with spectral properties similar to the parent compound. The same fluorescent properties and similar applications as FLUO-3, but has a lower affinity for calcium, and is insensitive to magnesium.

Other Notes

Cell permeable probe for fluorescence techniques: it has a lower affinity to calcium than Fluo-3 and no affinity to magnesium; abs.: λmax 462 nm; em.: λmax526 nm

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?