EMAIL THIS PAGE TO A FRIEND
17081 Sigma-Aldrich

Fura-2 LeakRes (AM)

for fluorescence, ≥95% (TLC)

Synonym: Fura PE 3-AM

Purchase

Properties

Related Categories Biochemicals and Reagents, Calcium Indicators, Chelators and Ion Probes, Fluorescent Indicators and Probes, Fluorescent Probes, Labels, Particles and Stains,
grade   for fluorescence
InChI Key   CGAZNAYDEAJZHQ-UHFFFAOYSA-N
assay   ≥95% (TLC)
solubility   DMSO: soluble
fluorescence   λex 335 nm; λem 495 nm in 0.1 M Tris pH 8.0, esterase; 10 mM Ca2+
storage temp.   −20°C

Description

Application

Fura 2 is one of the most widely used calcium indicators for ratiometric measurement and is highly suitable for digital imaging microscopy.

Features and Benefits

Pale yellow gum. Cell permeable acetoxymethyl ester (AM) derivative of Fura-PE3 with spectral properties similar to the parent compound.

Other Notes

Cell permeable ester of fluorescent calcium probe; abs.: λmax 364 nm; em.: λmax 502 nm

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

F0918 Fura-2 LeakRes (AM), ≥85%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?