• USA Home
  • 17084 - Indo PE 3 hexapotassium salt

EMAIL THIS PAGE TO A FRIEND
17084 Sigma-Aldrich

Indo PE 3 hexapotassium salt

for fluorescence, ≥95% (TLC)

  • Empirical Formula (Hill Notation) C40H37K6N5O15

  • Molecular Weight 1062.34

  •  MDL number MFCD01867221

  •  PubChem Substance ID 24850327

Purchase

Properties

Related Categories Biochemicals and Reagents, Calcium Indicators, Chelators and Ion Probes, Fluorescent Indicators and Probes, Fluorescent Probes, Labels, Particles and Stains,
grade   for fluorescence
InChI Key   GZHSNVHMGVZCAV-UHFFFAOYSA-H
assay   ≥95% (TLC)
solubility   H2O: soluble
fluorescence   λex 350 nm; λem 399 nm in 0.1 M Tris pH 8.0, 10 mM Ca2+
storage temp.   −20°C

Description

Application

Ratiomeric dye having similar spectral properties as Indo-1, but is leakage resistant

Features and Benefits

Yellow gum. A ratiometric dye with the same fluorescent properties and similar applications as Indo-1, but is leakage resistant.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?