EMAIL THIS PAGE TO A FRIEND
17086 Sigma-Aldrich

Indo PE 3-AM

for fluorescence, ≥90% (TLC)

  • Empirical Formula (Hill Notation) C58H67N5O27

  • Molecular Weight 1266.17

  •  MDL number MFCD01867222

  •  PubChem Substance ID 329751477

Purchase

Properties

Related Categories Biochemicals and Reagents, Calcium Indicators, Chelators and Ion Probes, Fluorescent Indicators and Probes, Fluorescent Probes, Labels, Particles and Stains,
grade   for fluorescence
InChI Key   LCOJDLFVQNLLLJ-UHFFFAOYSA-N
assay   ≥90% (TLC)
solubility   DMSO: soluble
fluorescence   λex 350 nm; λem 408 nm in 0.1 M Tris pH 8.0, esterase; 10 mM Ca2+
storage temp.   −20°C

Description

Application

Yellow gum. Cell permeable acetoxymethyl ester (AM) derivative of Indo-PE3 with spectral properties similar to the parent compound. A ratiometric dye with the same fluorescent properties and similar applications as Indo-1, but is leakage resistant.

Features and Benefits

Cell permeable ratiomeric dye having similar spectral properties as Indo-1, but is leakage resistant

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?