• USA Home
  • 17177 - 2-(Dodecyloxy)benzonitrile

EMAIL THIS PAGE TO A FRIEND
17177 Sigma-Aldrich

2-(Dodecyloxy)benzonitrile

Selectophore, ≥97.0%

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Plasticizer for ISE More...
grade   Selectophore
InChI Key   DDUJCCRSCBUHMI-UHFFFAOYSA-N
assay   ≥97.0% (GC)
  ≥97.0%
refractive index   n20/D 1.498
density   0.938 g/mL at 20 °C(lit.)

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Additive employed in a pH-insensitive potassium selective optode

Application

2-Cyanophenyl dodecyl ether (CPDDE) was used in preparing lipophilized gold nanoparticles which enhances optical sensors.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?