• USA Home
  • 17184 - m-HD Endo Broth with Brilliant Green

EMAIL THIS PAGE TO A FRIEND
17184 Sigma-Aldrich

m-HD Endo Broth with Brilliant Green

for microbiology

Synonym: Membrane filter HD Endo Broth with Brilliant Green, m-Hajna Damon Endo Broth with Brilliant Green

Purchase

Properties

Related Categories A - M, All Liquid Media/Broths, Escherichia coli & Coliforms, Media, Media by Application,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   brilliant green, 0.14 g/L
  casein enzymic hydrolysate, 10 g/L
  dipotassium phosphate, 6 g/L
  lactose, 20 g/L
  peptic digest of animal tissue, 10 g/L
  sodium chloride, 5 g/L
  sodium deoxycholate, 0.2 g/L
  sodium sulfite, 2.1 g/L
  yeast extract, 3 g/L
final pH   7.5±0.2 (25 °C)
suitability   selective for Escherichia coli and Coliforms

Description

Application

For the detection of coliforms in highly polluted waters using membrane filter technique.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?