• USA Home
  • 17300 - 2-Bromohexadecanoic acid

EMAIL THIS PAGE TO A FRIEND
17300 Sigma-Aldrich

2-Bromohexadecanoic acid

purum, ≥97.0% (GC)

Synonym: 2-Bromopalmitic acid

  • CAS Number 18263-25-7

  • Linear Formula CH3(CH2)13CH(Br)CO2H

  • Molecular Weight 335.32

  •  Beilstein Registry Number 1726517

  •  EC Number 242-137-0

  •  MDL number MFCD00004215

  •  PubChem Substance ID 329751526

Purchase

Properties

Related Categories Building Blocks, C13 to C42+, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   DPRAYRYQQAXQPE-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
mp   51-53 °C
solubility   methanol: soluble1 g/10 mL, clear, colorless

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of α-Bromopalmitic Acid Enantiomers on Astec® CHIROBIOTIC® T2

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of α-Bromopalmitic Acid Enantiomers on Astec® CHIROBIOTIC® T2
Keywords: Chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

238422 2-Bromohexadecanoic acid, ~97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?