EMAIL THIS PAGE TO A FRIEND
17341 Sigma-Aldrich

6-Bromo-1-hexene

technical, ≥90% (GC)

Purchase

Properties

Related Categories Alkenyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   RIMXEJYJXDBLIE-UHFFFAOYSA-N
grade   technical
assay   ≥90% (GC)
contains   silver wool as stabilizer
impurities   6-bromo-2-hexene (cis+trans)
refractive index   n20/D 1.465(lit.)
  n20/D 1.466
bp   47-51 °C/16 mmHg(lit.)
density   1.22 g/mL at 25 °C(lit.)

Description

Application

6-Bromo-1-hexene was used to evaluate the effectiveness of immobilization of catalytic palladium nanoparticles in microfluidic reactors. It was also used in the sequential isomerization/styrylation reactions of cyclopropylmethyl bromide.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
129.2 °F
Flash Point(C) 
54 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

247219 6-Bromo-1-hexene, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?