• USA Home
  • 17399 - (S)-(+)-1-Benzyloxy-2-propanol

EMAIL THIS PAGE TO A FRIEND
17399 Sigma-Aldrich

(S)-(+)-1-Benzyloxy-2-propanol

Synonym: (S)-1-O-Benzylpropylene glycol

  • CAS Number 85483-97-2

  • Empirical Formula (Hill Notation) C10H14O2

  • Molecular Weight 166.22

  •  MDL number MFCD07367008

Purchase

Properties

InChI Key   KJBPYIUAQLPHJG-VIFPVBQESA-N
refractive index   n20/D 1.510(lit.)
density   1.044 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?