• USA Home
  • 17431 - α-Bromoisobutyric acid

EMAIL THIS PAGE TO A FRIEND
17431 Sigma-Aldrich

α-Bromoisobutyric acid

purum, ≥98.0% (GC)

Synonym: 2-Bromo-2-methylpropionic acid

  • CAS Number 2052-01-9

  • Linear Formula (CH3)2CBrCOOH

  • Molecular Weight 167.00

  •  Beilstein Registry Number 1745543

  •  EC Number 218-139-2

  •  MDL number MFCD00004168

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   XXSPGBOGLXKMDU-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   198-200 °C(lit.)
mp   44-47 °C(lit.)
solubility   alcohol: soluble
  cold water: very slightly soluble
  diethyl ether: soluble

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

α-Bromoisobutyric acid (2-Bromo-2-methylpropionic acid) was used in the preparation of multifunctional silica colloids. It was also used to develop tumor specific, water dispersible superparamagnetic iron oxide nanoparticles (SPIONs).

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

306851 2-Bromo-2-methylpropionic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?