• USA Home
  • 17440 - Ethyl α-bromoisobutyrate

EMAIL THIS PAGE TO A FRIEND
17440 Sigma-Aldrich

Ethyl α-bromoisobutyrate

purum, ≥97.0% (GC)

Synonym: Ethyl 2-bromo-2-methylpropionate

  • CAS Number 600-00-0

  • Linear Formula (CH3)2CBrCOOC2H5

  • Molecular Weight 195.05

  •  Beilstein Registry Number 1098947

  •  EC Number 209-980-6

  •  MDL number MFCD00000123

  •  PubChem Substance ID 24850480

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   IOLQWGVDEFWYNP-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.444(lit.)
  n20/D 1.445
bp   65-67 °C/11 mmHg(lit.)
density   1.329 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Ethyl α-bromoisobutyrate was used in the synthesis of ketals and ulosonic acid esters.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
140 °F
Flash Point(C) 
60 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?