• USA Home
  • 17460 - α-Bromoisovaleric acid

EMAIL THIS PAGE TO A FRIEND
17460 Sigma-Aldrich

α-Bromoisovaleric acid

purum, ≥98.0% (GC)

Synonym: 2-Bromoisovaleric acid

  • CAS Number 565-74-2

  • Linear Formula (CH3)2CHCHBrCOOH

  • Molecular Weight 181.03

  •  Beilstein Registry Number 1721146

  •  EC Number 209-291-0

  •  MDL number MFCD00004210

  •  PubChem Substance ID 24850487

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   UEBARDWJXBGYEJ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   124-126 °C/20 mmHg(lit.)
mp   39-42 °C(lit.)
  39-43 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

α-Bromoisovaleric acid was used to study the conjugation of glutathione (GSH) with (R)-and (S)-α-bromoisovaleric acid (BI) in the rat.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
RTECS 
ES8055000
Flash Point(F) 
224.6 °F
Flash Point(C) 
107 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?