• USA Home
  • 17540 - 2-Bromo-6-methoxynaphthalene

EMAIL THIS PAGE TO A FRIEND
17540 Sigma-Aldrich

2-Bromo-6-methoxynaphthalene

purum, ≥98.0% (HPLC)

Purchase

Properties

Related Categories Building Blocks, C11, Chemical Synthesis, Ethers, Organic Building Blocks,
InChI Key   AYFJBMBVXWNYLT-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   106-109 °C(lit.)
  106-109 °C

Description

Application

2-Bromo-6-methoxynaphthalene was used to study the catalytic activity of the trimetallic nanoparticles.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?