• USA Home
  • 17560 - 2-Bromomethyl-anthraquinone

EMAIL THIS PAGE TO A FRIEND
17560 Sigma-Aldrich

2-Bromomethyl-anthraquinone

purum, ≥97.0% (HPLC)

Synonym: MAQ-Br

  • CAS Number 7598-10-9

  • Empirical Formula (Hill Notation) C15H9BrO2

  • Molecular Weight 301.13

  •  Beilstein Registry Number 2276854

  •  MDL number MFCD00042958

  •  PubChem Substance ID 24850514

Purchase

Properties

Related Categories Building Blocks, C15 to C38, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   FCETXLHFBYOVCV-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (HPLC)
impurities   ≤0.5% water
mp   203 °C (dec.)(lit.)

Description

Other Notes

Reagent for introducing the MAQ protecting group

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?