• USA Home
  • 17568 - 3-(Bromomethyl)benzonitrile

EMAIL THIS PAGE TO A FRIEND
17568 Sigma-Aldrich

3-(Bromomethyl)benzonitrile

purum, ≥96.0% (HPLC)

Synonym: α-Bromo-m-tolunitrile, 3-Cyanobenzyl bromide

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
InChI Key   CVKOOKPNCVYHNY-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (HPLC)
mp   91-93 °C

Description

Application

3-(Bromomethyl)benzonitrile (-Bromo-m-tolunitrile) was used in the synthesis of 3-[(Pyrazol-1-ylmethyl)]benzonitrile. It was also used in the alkylation of pyrazole for the synthesis of carbonitrile derivative.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

145610 3-(Bromomethyl)benzonitrile, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?