• USA Home
  • 17570 - 4-(Bromomethyl)benzonitrile

EMAIL THIS PAGE TO A FRIEND
17570 Sigma-Aldrich

4-(Bromomethyl)benzonitrile

purum, ≥95.0% (HPLC)

Synonym: α-Bromo-p-tolunitrile, 4-Cyanobenzyl bromide

  • CAS Number 17201-43-3

  • Linear Formula BrCH2C6H4CN

  • Molecular Weight 196.04

  •  Beilstein Registry Number 636717

  •  EC Number 241-246-0

  •  MDL number MFCD00001829

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
InChI Key   UMLFTCYAQPPZER-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (HPLC)
mp   114-117 °C
  115-117 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

144061 4-(Bromomethyl)benzonitrile, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?