• USA Home
  • 17619 - (R)-(−)-3-Bromo-2-methyl-1-propanol

EMAIL THIS PAGE TO A FRIEND
17619 Sigma-Aldrich

(R)-(−)-3-Bromo-2-methyl-1-propanol

purum, ≥96.0% (sum of enantiomers, GC)

  • CAS Number 93381-28-3

  • Linear Formula BrCH2CH(CH3)CH2OH

  • Molecular Weight 153.02

  •  Beilstein Registry Number 3600269

  •  MDL number MFCD00013302

  •  PubChem Substance ID 329751549

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   KIBOHRIGZMLNNS-BYPYZUCNSA-N
grade   purum
assay   ≥96.0% (sum of enantiomers, GC)
optical activity   [α]20/D −6.5±1°, c = 2% in chloroform
refractive index   n20/D 1.482
  n20/D 1.484(lit.)
bp   73-74 °C/9 mmHg(lit.)
density   1.461 g/mL at 20 °C(lit.)

Description

Other Notes

Chiral building block for various alkylation and coupling reactions

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
186.8 °F
Flash Point(C) 
86 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?