• USA Home
  • 17620 - (S)-(+)-3-Bromo-2-methyl-1-propanol

EMAIL THIS PAGE TO A FRIEND
17620 Sigma-Aldrich

(S)-(+)-3-Bromo-2-methyl-1-propanol

purum, ≥96.0% (sum of enantiomers, GC)

  • CAS Number 98244-48-5

  • Linear Formula BrCH2CH(CH3)CH2OH

  • Molecular Weight 153.02

  •  Beilstein Registry Number 4738280

  •  MDL number MFCD00064575

  •  PubChem Substance ID 329751550

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   KIBOHRIGZMLNNS-SCSAIBSYSA-N
grade   purum
assay   ≥96.0% (sum of enantiomers, GC)
optical activity   [α]20/D +6.5±1°, c = 2% in chloroform
refractive index   n20/D 1.483(lit.)
  n20/D 1.483
density   1.461 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
197.6 °F
Flash Point(C) 
92 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?