• USA Home
  • 17623 - 4-Bromomethyl-7-methoxycoumarin

EMAIL THIS PAGE TO A FRIEND
17623 Sigma-Aldrich

4-Bromomethyl-7-methoxycoumarin

for fluorescence, ≥97.0% (HPLC)

Synonym: BMC, Br-Mmc

  • CAS Number 35231-44-8

  • Empirical Formula (Hill Notation) C11H9BrO3

  • Molecular Weight 269.09

  •  Beilstein Registry Number 187211

  •  EC Number 252-448-3

  •  MDL number MFCD00006869

  •  PubChem Substance ID 24850565

Purchase

Description

Application

Fluorescent label for fatty acids.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3335
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

235202 4-Bromomethyl-7-methoxycoumarin, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?