EMAIL THIS PAGE TO A FRIEND
17740 Sigma-Aldrich

1-Bromononane

purum, ≥97.0% (GC)

Synonym: Nonyl bromide

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   AYMUQTNXKPEMLM-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.454(lit.)
  n20/D 1.454
bp   201 °C(lit.)
density   1.084 g/mL at 25 °C(lit.)

Description

Application

1-Bromononane was used to quench dianion to synthesize alcohol.

General description

1-Bromononane undergoes degradation on refluxing with ethanolic potassium hydroxide.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
194 °F
Flash Point(C) 
90 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

B74607 1-Bromononane, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?