EMAIL THIS PAGE TO A FRIEND
17743 Sigma-Aldrich

1-Bromododecane

puriss., ≥99.5% (GC)

Synonym: Dodecyl bromide, Lauryl bromide

  • CAS Number 143-15-7

  • Linear Formula CH3(CH2)11Br

  • Molecular Weight 249.23

  •  Beilstein Registry Number 506159

  •  EC Number 205-587-9

  •  MDL number MFCD00000225

  •  PubChem Substance ID 24850636

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
vapor density   8.6 (vs air)
InChI Key   PBLNBZIONSLZBU-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.5% (GC)
refractive index   n20/D 1.458(lit.)
  n20/D 1.458
bp   134-135 °C/6 mmHg(lit.)
mp   −11-−9 °C(lit.)
solubility   H2O: insoluble
  alcohol: soluble
  diethyl ether: soluble
density   1.040 g/mL at 20 °C
  1.038 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

1-Bromododecane was used in the synthesis of (−)-muricatacin. It was also used in the synthesis of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?