EMAIL THIS PAGE TO A FRIEND
17770 Sigma-Aldrich

1-Bromooctadecane

purum, ≥97.0% (GC)

Synonym: Octadecyl bromide, Stearyl bromide

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   WSULSMOGMLRGKU-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
bp   214-216 °C/12 mmHg(lit.)
mp   25-30 °C(lit.)
  25-30 °C
density   0.976 g/mL at 25 °C(lit.)

Description

Application

1-Bromooctadecane was used in the synthesis of shortened single-walled carbon nanotubes.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

199494 1-Bromooctadecane, ≥97.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?