• USA Home
  • 17871 - C-Phycocyanin, activated

EMAIL THIS PAGE TO A FRIEND
17871 Sigma-Aldrich

C-Phycocyanin, activated

Synonym: Activated C-Phycocyanin

Purchase

Description

Reconstitution

C-Phycocyanin was treated with a 15 molar excess of succinimidyl-5-acetylthioacetate. Reconstitute to 300 μl with distilled or deionized water (the solution will consist of 100 mM sodium phosphate pH 7.2, 50 mM sucrose, 0.05 % sodium azide and 5 mg/ml pigment).
Do not freeze, store at 4 °C in the dark.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?